Assay Design
Gene Transcripts
- Gene Symbol
- Podnl1
- Entrez Gene ID
-
288907
- Gene Name
- podocan-like 1
- Gene Aliases
-
RGD1307753
- Chromosome Location
-
Chr.19: 25240089 - 25246583 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_213845.7 |
XP_213845.6 |
9 - 10 |
1665 |
57 |
View Full Product Details