Assay Design



Gene Transcripts

Gene Symbol
Wdr83
Entrez Gene ID
288924
Gene Name
WD repeat domain 83
Gene Aliases
Morg1, RGD1306947
Chromosome Location
Chr.19: 26188644 - 26194193 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047847.1 NP_001041312.1 6 - 7 640 82
GenBank mRNA AY940050.1 - 6 - 7 640 82

View Full Product Details