Assay Design



Gene Transcripts

Gene Symbol
Otud4
Entrez Gene ID
307774
Gene Name
OTU deubiquitinase 4
Gene Aliases
RGD1305606
Chromosome Location
Chr.19: 31895753 - 31942516 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191700.1 NP_001178629.1 12 - 13 1287 88
XM_006255402.2 XP_006255464.1 12 - 13 1626 88
XM_008772493.1 XP_008770715.1 12 - 13 1802 88
XM_008772494.1 XP_008770716.1 12 - 13 1358 88
XM_008772495.1 XP_008770717.1 13 - 14 1406 88

View Full Product Details