Assay Design



Gene Transcripts

Gene Symbol
Slco6c1
Entrez Gene ID
287006
Gene Name
solute carrier organic anion transporter family, member 6c1
Gene Aliases
GST-2
Chromosome Location
Chr.9: 104622219 - 104700657 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173338.1 NP_775460.1 - 1022 100
XM_008767361.1 XP_008765583.1 - 904 100
GenBank mRNA AF326113.1 - - 1022 100

View Full Product Details