Assay Design



Gene Transcripts

Gene Symbol
Neu4
Entrez Gene ID
316642
Gene Name
sialidase 4
Gene Aliases
-
Chromosome Location
Chr.9: 101284902 - 101290558 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108234.1 NP_001101704.1 - 629 81
XM_008767376.1 XP_008765598.1 - 1989 81

Gene Symbol
NEWGENE_1308624
Entrez Gene ID
103690047
Gene Name
sialidase 4
Gene Aliases
Neu4
Chromosome Location
Chr.9: 101003469 - 101008365 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006245535.2 XP_006245597.1 2 - 3 1989 81
XM_008767394.1 XP_008765616.1 3 - 4 610 81

View Full Product Details