Assay Design



Gene Transcripts

Gene Symbol
Itgb2
Entrez Gene ID
309684
Gene Name
integrin, beta 2
Gene Aliases
Cd18
Chromosome Location
Chr.20: 11777773 - 11815647 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037780.2 NP_001032869.2 14 - 15 2331 142
GenBank mRNA BC099151.1 - 3 - 4 1162 142
BC166721.1 - 14 - 15 2310 142

View Full Product Details