Assay Design



Gene Transcripts

Gene Symbol
Itgb2
Entrez Gene ID
309684
Gene Name
integrin, beta 2
Gene Aliases
Cd18
Chromosome Location
Chr.20: 11777773 - 11815647 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037780.2 NP_001032869.2 6 - 7 995 68
GenBank mRNA BC166721.1 - 6 - 7 974 68

View Full Product Details