Assay Design



Gene Transcripts

Gene Symbol
Wdr4
Entrez Gene ID
690032
Gene Name
WD repeat domain 4
Gene Aliases
-
Chromosome Location
Chr.20: 10240571 - 10264815 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135667.1 NP_001129139.1 1 - 2 136 67
XM_008772828.1 XP_008771050.1 1 - 2 347 67

View Full Product Details