Assay Design



Gene Transcripts

Gene Symbol
Ccdc6
Entrez Gene ID
691155
Gene Name
coiled-coil domain containing 6
Gene Aliases
-
Chromosome Location
Chr.20: 19729013 - 19825245 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001077030.3 XP_001077030.1 7 - 8 1182 76

View Full Product Details