Assay Design



Gene Transcripts

Gene Symbol
Umodl1
Entrez Gene ID
365544
Gene Name
uromodulin-like 1
Gene Aliases
RGD1566039
Chromosome Location
Chr.20: 9643509 - 9703091 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008772853.1 XP_008771075.1 13 - 14 2606 65

View Full Product Details