Assay Design
Gene Transcripts
- Gene Symbol
- Tff1
- Entrez Gene ID
-
117270
- Gene Name
- trefoil factor 1
- Gene Aliases
-
Ps2
- Chromosome Location
-
Chr.20: 9892124 - 9895984 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057129.1 |
NP_476470.1 |
2 - 3 |
257 |
131 |
GenBank mRNA |
D83231.1 |
- |
2 - 3 |
257 |
131 |
View Full Product Details