Assay Design



Gene Transcripts

Gene Symbol
Tmprss3
Entrez Gene ID
309665
Gene Name
transmembrane protease, serine 3
Gene Aliases
-
Chromosome Location
Chr.20: 9910522 - 9930187 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107619.1 NP_001101089.1 9 - 10 1102 111

View Full Product Details