Assay Design



Gene Transcripts

Gene Symbol
Pofut2
Entrez Gene ID
309686
Gene Name
protein O-fucosyltransferase 2
Gene Aliases
-
Chromosome Location
Chr.20: 12117811 - 12129342 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107621.2 NP_001101091.2 6 - 7 899 87
XM_006256299.2 XP_006256361.1 6 - 7 1494 87
GenBank mRNA DY565624.1 - 2 - 3 165 87

View Full Product Details