Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008884.2 | NP_001008884.1 | 5 - 6 | 845 | 81 |
GenBank mRNA | AF084933.1 | - | 5 - 6 | 785 | 81 |
BC087691.1 | - | 6 - 7 | 765 | 81 | |
CV111238.1 | - | 1 - 2 | 81 | ||
FQ225193.1 | - | 5 - 6 | 833 | 81 | |
FQ226724.1 | - | 5 - 6 | 833 | 81 | |
FQ226850.1 | - | 5 - 6 | 833 | 81 | |
FQ227087.1 | - | 5 - 6 | 830 | 81 | |
FQ228005.1 | - | 5 - 6 | 833 | 81 | |
FQ228124.1 | - | 5 - 6 | 810 | 81 | |
FQ228129.1 | - | 5 - 6 | 833 | 81 | |
FQ228185.1 | - | 5 - 6 | 851 | 81 | |
FQ230360.1 | - | 5 - 6 | 833 | 81 | |
FQ231387.1 | - | 5 - 6 | 833 | 81 | |
FQ231709.1 | - | 5 - 6 | 833 | 81 | |
FQ232046.1 | - | 5 - 6 | 833 | 81 | |
FQ232100.1 | - | 5 - 6 | 832 | 81 | |
FQ232453.1 | - | 5 - 6 | 833 | 81 | |
FQ233049.1 | - | 5 - 6 | 829 | 81 | |
FQ233335.1 | - | 5 - 6 | 833 | 81 | |
FQ233396.1 | - | 5 - 6 | 833 | 81 | |
FQ233493.1 | - | 5 - 6 | 833 | 81 | |
FQ233800.1 | - | 5 - 6 | 811 | 81 | |
FQ233801.1 | - | 5 - 6 | 835 | 81 | |
FQ234464.1 | - | 5 - 6 | 832 | 81 | |
FQ234907.1 | - | 5 - 6 | 833 | 81 | |
X53054.1 | - | 5 - 6 | 796 | 81 |