Assay Design



Gene Transcripts

Gene Symbol
Lhfpl5
Entrez Gene ID
294303
Gene Name
lipoma HMGIC fusion partner-like 5
Gene Aliases
Tmhs
Chromosome Location
Chr.20: 5815837 - 5826137 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013906.1 NP_001013928.1 3 - 4 939 97
GenBank mRNA BC087673.1 - 3 - 4 939 97

View Full Product Details