Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109672.1 | NP_001103142.1 | 8 - 9 | 856 | 103 |
XM_006240549.2 | XP_006240611.1 | 9 - 10 | 1099 | 103 | |
XM_006240551.2 | XP_006240613.1 | 7 - 8 | 1101 | 103 | |
GenBank mRNA | BC129121.1 | - | 8 - 9 | 856 | 103 |