Assay Design



Gene Transcripts

Gene Symbol
Tnfaip2
Entrez Gene ID
299339
Gene Name
tumor necrosis factor, alpha-induced protein 2
Gene Aliases
-
Chromosome Location
Chr.6: 135890874 - 135903647 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001137633.1 NP_001131105.1 12 - 13 2213 70
XM_006240577.2 XP_006240639.1 11 - 12 2221 70
GenBank mRNA FQ212109.1 - 11 - 12 1971 70

View Full Product Details