Assay Design



Gene Transcripts

Gene Symbol
Wdr25l
Entrez Gene ID
314443
Gene Name
WD repeat domain 25-like
Gene Aliases
RGD1562505, Wdr25
Chromosome Location
Chr.6: 132802329 - 132932254 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135894.1 NP_001129366.1 1 - 2 95 79
XM_006240544.2 XP_006240606.1 2 - 3 312 79
XM_006240545.2 XP_006240607.1 2 - 3 157 79
XM_006240546.2 XP_006240608.1 2 - 3 175 79
XM_006240547.2 XP_006240609.1 2 - 3 383 79
GenBank mRNA BC166712.1 - 1 - 2 95 79

View Full Product Details