Assay Design
Gene Transcripts
- Gene Symbol
- Hhipl1
- Entrez Gene ID
-
362781
- Gene Name
- HHIP-like 1
- Gene Aliases
-
RGD1311698
- Chromosome Location
-
Chr.6: 132218593 - 132242852 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_343107.7 |
XP_343108.4 |
3 - 4 |
1558 |
77 |
View Full Product Details