Assay Design



Gene Transcripts

Gene Symbol
Rmnd1
Entrez Gene ID
292268
Gene Name
required for meiotic nuclear division 1 homolog (S. cerevisiae)
Gene Aliases
Iag-1, RGD1309546
Chromosome Location
Chr.1: 40948843 - 40982669 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006227834.1 XP_006227896.1 3 - 4 784 71
XM_006227837.2 XP_006227899.1 3 - 4 1301 71
GenBank mRNA FQ233028.1 - 2 - 3 171 71

View Full Product Details