Assay Design



Gene Transcripts

Gene Symbol
Tfb1m
Entrez Gene ID
308140
Gene Name
transcription factor B1, mitochondrial
Gene Aliases
-
Chromosome Location
Chr.1: 44433116 - 44474674 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181474.2 NP_852139.2 6 - 7 812 110
GenBank mRNA AY198392.1 - 6 - 7 792 110
BC070907.1 - 6 - 7 812 110

View Full Product Details