Assay Design



Gene Transcripts

Gene Symbol
Vip
Entrez Gene ID
117064
Gene Name
vasoactive intestinal peptide
Gene Aliases
vip/phi27
Chromosome Location
Chr.1: 42165043 - 42177582 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053991.1 NP_446443.1 4 - 5 485 126
XM_006227827.2 XP_006227889.1 5 - 6 930 126
GenBank mRNA AB089807.1 - 4 - 5 407 126
AB089808.1 - 3 - 4 642 126
AB089809.1 - 4 - 5 502 126
CB583853.1 - 4 - 5 499 126
X02341.1 - 3 - 4 320 126

View Full Product Details