Assay Design



Gene Transcripts

Gene Symbol
Tspan7
Entrez Gene ID
363447
Gene Name
tetraspanin 7
Gene Aliases
A15, DXS1692E, MXS1, TALLA-1, Tm4sf2
Chromosome Location
Chr.X: 13261551 - 13282886 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC079470.1 - 4 - 5 579 115
BC158792.1 - 4 - 5 620 115
EF688600.1 - 4 - 5 579 115

View Full Product Details