Assay Design



Gene Transcripts

Gene Symbol
Fundc1
Entrez Gene ID
363442
Gene Name
FUN14 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.X: 5549468 - 5566125 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025027.1 NP_001020198.1 1 - 2 161 79
GenBank mRNA BC091353.1 - 1 - 2 161 79

View Full Product Details