Assay Design



Gene Transcripts

Gene Symbol
Cfp
Entrez Gene ID
299314
Gene Name
complement factor properdin
Gene Aliases
Pfc
Chromosome Location
Chr.X: 1311121 - 1316683 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106757.2 NP_001100227.1 6 - 7 943 62
GenBank mRNA BC158889.1 - 6 - 7 943 62
FQ212442.1 - 6 - 7 978 62

View Full Product Details