Assay Design



Gene Transcripts

Gene Symbol
Cfp
Entrez Gene ID
299314
Gene Name
complement factor properdin
Gene Aliases
Pfc
Chromosome Location
Chr.X: 1311121 - 1316683 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106757.2 NP_001100227.1 8 - 9 1252 119
GenBank mRNA BC158889.1 - 8 - 9 1252 119
FQ209917.1 - 8 - 9 1285 119
FQ212442.1 - 8 - 9 1287 119

View Full Product Details