Assay Design



Gene Transcripts

Gene Symbol
Slc10a7
Entrez Gene ID
291942
Gene Name
solute carrier family 10, member 7
Gene Aliases
P7
Chromosome Location
Chr.19: 32857984 - 33080772 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001010948.1 NP_001010948.1 7 - 8 551 98
GenBank mRNA AY825929.1 - 7 - 8 551 98
FQ211514.1 - - 743 98

View Full Product Details