Assay Design



Gene Transcripts

Gene Symbol
Cdk10
Entrez Gene ID
361434
Gene Name
cyclin-dependent kinase 10
Gene Aliases
-
Chromosome Location
Chr.19: 56024903 - 56032610 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025722.2 NP_001020893.2 9 - 10 1254 99
NM_001109936.1 NP_001103406.1 10 - 11 803 99
NM_001109937.1 NP_001103407.1 10 - 11 764 99
GenBank mRNA BC098804.1 - 9 - 10 1254 99

View Full Product Details