Assay Design



Gene Transcripts

Gene Symbol
Cmc2
Entrez Gene ID
100363376
Gene Name
C-x(9)-C motif containing 2
Gene Aliases
-
Chromosome Location
Chr.19: 49433332 - 49457633 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002728617.4 XP_002728663.1 3 - 4 283 61
XM_002728618.3 XP_002728664.1 2 - 3 314 61
XM_006255698.1 XP_006255760.1 3 - 4 312 61
XM_006255699.2 XP_006255761.1 3 - 4 254 61
GenBank mRNA FQ229378.1 - 3 - 4 533 61

View Full Product Details