Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100922.1 | NP_001094392.1 | 8 - 9 | 907 | 94 |
XM_006255219.1 | XP_006255281.1 | 8 - 9 | 841 | 94 | |
GenBank mRNA | AY375243.1 | - | 8 - 9 | 753 | 94 |
BC129128.1 | - | 8 - 9 | 907 | 94 | |
X57986.1 | - | 8 - 9 | 817 | 94 |