Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017511.1 | NP_001017511.1 | 4 - 5 | 1401 | 118 |
XM_006255718.2 | XP_006255780.1 | 4 - 5 | 1513 | 118 | |
XM_008772641.1 | XP_008770863.1 | 4 - 5 | 2763 | 118 | |
GenBank mRNA | BC081914.1 | - | 4 - 5 | 1401 | 118 |