Assay Design



Gene Transcripts

Gene Symbol
C8a
Entrez Gene ID
298288
Gene Name
complement component 8, alpha polypeptide
Gene Aliases
-
Chromosome Location
Chr.5: 124348536 - 124403117 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106670.1 NP_001100140.1 8 - 9 1258 110
GenBank mRNA FQ211202.1 - 8 - 9 1337 110
FQ211244.1 - 8 - 9 1337 110

View Full Product Details