Assay Design



Gene Transcripts

Gene Symbol
Atg4c
Entrez Gene ID
313391
Gene Name
autophagy related 4C, cysteine peptidase
Gene Aliases
-
Chromosome Location
Chr.5: 117862746 - 117931520 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107948.1 NP_001101418.1 2 - 3 516 146

View Full Product Details