Assay Design



Gene Transcripts

Gene Symbol
Sh3rf3
Entrez Gene ID
294557
Gene Name
SH3 domain containing ring finger 3
Gene Aliases
Sh3md4
Chromosome Location
Chr.20: 28478636 - 28720020 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008772954.1 XP_008771176.1 9 - 10 1977 100

View Full Product Details