Assay Design



Gene Transcripts

Gene Symbol
Micu1
Entrez Gene ID
365567
Gene Name
mitochondrial calcium uptake 1
Gene Aliases
Cbara1
Chromosome Location
Chr.20: 29284848 - 29433617 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199412.1 NP_955444.1 8 - 9 977 74
XM_006256420.2 XP_006256482.1 8 - 9 1092 74
XM_006256421.2 XP_006256483.1 9 - 10 1085 74
XM_006256422.2 XP_006256484.1 9 - 10 1079 74
GenBank mRNA BC062075.1 - 8 - 9 977 74

View Full Product Details