Assay Design



Gene Transcripts

Gene Symbol
Mcu
Entrez Gene ID
294560
Gene Name
mitochondrial calcium uniporter
Gene Aliases
Ccdc109a
Chromosome Location
Chr.20: 29038906 - 29199224 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106398.1 NP_001099868.1 2 - 3 227 77

View Full Product Details