Assay Design
Gene Transcripts
- Gene Symbol
- Tacr2
- Entrez Gene ID
-
25007
- Gene Name
- tachykinin receptor 2
- Gene Aliases
-
Tac2r
- Chromosome Location
-
Chr.20: 31892515 - 31905153 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080768.1 |
NP_542946.1 |
3 - 4 |
1183 |
68 |
GenBank mRNA |
M31838.1 |
- |
3 - 4 |
1183 |
68 |
View Full Product Details