Assay Design



Gene Transcripts

Gene Symbol
P4ha1
Entrez Gene ID
64475
Gene Name
prolyl 4-hydroxylase, alpha polypeptide I
Gene Aliases
PHalphaI
Chromosome Location
Chr.20: 28920616 - 28972576 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172062.2 NP_742059.2 3 - 4 413 101
XM_006256424.2 XP_006256486.1 4 - 5 498 101
XM_006256425.2 XP_006256487.1 3 - 4 407 101
GenBank mRNA BC078703.1 - 3 - 4 413 101
X78949.1 - 3 - 4 247 101

View Full Product Details