Assay Design



Gene Transcripts

Gene Symbol
Tmem17
Entrez Gene ID
360985
Gene Name
transmembrane protein 17
Gene Aliases
-
Chromosome Location
Chr.14: 107302808 - 107308226 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001010961.1 NP_001010961.1 3 - 4 662 78
GenBank mRNA BC089059.1 - 3 - 4 662 78
FQ228748.1 - 3 - 4 320 78

View Full Product Details