Assay Design



Gene Transcripts

Gene Symbol
Ugp2
Entrez Gene ID
289827
Gene Name
UDP-glucose pyrophosphorylase 2
Gene Aliases
-
Chromosome Location
Chr.14: 106208174 - 106249322 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024743.1 NP_001019914.1 1 - 2 262 115
GenBank mRNA BC097369.1 - 1 - 2 262 115

View Full Product Details