Assay Design



Gene Transcripts

Gene Symbol
Cd83
Entrez Gene ID
361226
Gene Name
CD83 molecule
Gene Aliases
-
Chromosome Location
Chr.17: 24416651 - 24435790 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108410.1 NP_001101880.1 2 - 3 261 56
GenBank mRNA BC166582.1 - 2 - 3 172 56

View Full Product Details