Assay Design



Gene Transcripts

Gene Symbol
Mylip
Entrez Gene ID
306825
Gene Name
myosin regulatory light chain interacting protein
Gene Aliases
MIR
Chromosome Location
Chr.17: 19682040 - 19703681 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107344.2 NP_001100814.2 4 - 5 881 77

View Full Product Details