Assay Design



Gene Transcripts

Gene Symbol
Rabepk
Entrez Gene ID
296649
Gene Name
Rab9 effector protein with kelch motifs
Gene Aliases
9530020d24rik, RGD1310612
Chromosome Location
Chr.3: 13845352 - 13865843 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024871.1 NP_001020042.1 6 - 7 955 78
XM_006233955.1 XP_006234017.1 5 - 6 1002 78
XM_006233957.2 XP_006234019.1 5 - 6 610 78
XM_006233958.1 XP_006234020.1 5 - 6 869 78
XM_006233959.2 XP_006234021.1 5 - 6 677 78
XM_006233963.1 XP_006234025.1 5 - 6 918 78
XM_006233964.2 XP_006234026.1 5 - 6 527 78
XM_006233966.1 XP_006234028.1 5 - 6 914 78
XM_006233967.1 XP_006234029.1 5 - 6 781 78
XM_008761724.1 XP_008759946.1 7 - 8 1152 78
XM_008761725.1 XP_008759947.1 6 - 7 707 78
XM_008761726.1 XP_008759948.1 6 - 7 683 78
XM_008761727.1 XP_008759949.1 5 - 6 1029 78
XM_008761728.1 XP_008759950.1 5 - 6 638 78
XM_008761729.1 XP_008759951.1 5 - 6 896 78
XM_008761730.1 XP_008759952.1 6 - 7 954 78
XM_008761731.1 XP_008759953.1 6 - 7 599 78
XM_008761732.1 XP_008759954.1 6 - 7 596 78
XM_008761733.1 XP_008759955.1 4 - 5 920 78
XM_008761734.1 XP_008759956.1 5 - 6 557 78
XM_008761735.1 XP_008759957.1 5 - 6 554 78
XM_008761736.1 XP_008759958.1 4 - 5 572 78
XM_008761737.1 XP_008759959.1 5 - 6 530 78
XM_008761738.1 XP_008759960.1 5 - 6 785 78
XM_008761739.1 XP_008759961.1 4 - 5 831 78
GenBank mRNA BC097415.1 - 6 - 7 955 78

View Full Product Details