Assay Design
Gene Transcripts
- Gene Symbol
- Gpsm1
- Entrez Gene ID
-
246254
- Gene Name
- G-protein signaling modulator 1
- Gene Aliases
-
Ags3
- Chromosome Location
-
Chr.3: 3767394 - 3794360 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_144745.2 |
NP_653346.2 |
1 - 2 |
188 |
113 |
GenBank mRNA |
CB743977.1 |
- |
1 - 2 |
236 |
113 |
View Full Product Details