Assay Design



Gene Transcripts

Gene Symbol
Fam73b
Entrez Gene ID
296623
Gene Name
family with sequence similarity 73, member B
Gene Aliases
RGD1310887
Chromosome Location
Chr.3: 8928495 - 8951189 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106566.1 NP_001100036.1 5 - 6 727 59
XM_006233781.1 XP_006233843.1 5 - 6 776 59
XM_006233782.2 XP_006233844.1 5 - 6 792 59
GenBank mRNA FQ212119.1 - 5 - 6 695 59
FQ231217.1 - 5 - 6 731 59

View Full Product Details