Assay Design



Gene Transcripts

Gene Symbol
Fam73b
Entrez Gene ID
296623
Gene Name
family with sequence similarity 73, member B
Gene Aliases
RGD1310887
Chromosome Location
Chr.3: 8928495 - 8951189 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106566.1 NP_001100036.1 14 - 15 1652 73

View Full Product Details