Assay Design



Gene Transcripts

Gene Symbol
Qsox2
Entrez Gene ID
681023
Gene Name
quiescin Q6 sulfhydryl oxidase 2
Gene Aliases
-
Chromosome Location
Chr.3: 3662324 - 3691972 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109434.1 NP_001102904.1 6 - 7 859 125

View Full Product Details