Assay Design



Gene Transcripts

Gene Symbol
Rexo4
Entrez Gene ID
311826
Gene Name
REX4, RNA exonuclease 4 homolog (S. cerevisiae)
Gene Aliases
Xpmc2h
Chromosome Location
Chr.3: 5500578 - 5510908 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033884.1 NP_001029056.1 1 - 2 279 117
GenBank mRNA BC161820.1 - 1 - 2 244 117
CO563573.1 - - 279 117

View Full Product Details