Assay Design



Gene Transcripts

Gene Symbol
Lcn9
Entrez Gene ID
296578
Gene Name
lipocalin 9
Gene Aliases
RGD1306341
Chromosome Location
Chr.3: 3272173 - 3274715 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106560.1 NP_001100030.1 2 - 3 244 90
XM_008761622.1 XP_008759844.1 2 - 3 264 90
GenBank mRNA DQ537496.1 - 2 - 3 233 90

View Full Product Details