Assay Design



Gene Transcripts

Gene Symbol
Tbpl2
Entrez Gene ID
680050
Gene Name
TATA box binding protein-like 2
Gene Aliases
-
Chromosome Location
Chr.3: 1095883 - 1116677 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099361.1 NP_001092831.1 5 - 6 868 165

View Full Product Details